missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Adiponectin/Acrp30 Antibody, R&D Systems™

Goat Polyclonal Antibody
Marca: R&D Systems AF1065-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Adiponectin/Acrp30 Polyclonal specifically detects Adiponectin/Acrp30 in Human samples. It is validated for Western Blot, Simple Western.
Especificaciones
| Adiponectin/Acrp30 | |
| Polyclonal | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL | |
| Q15848 | |
| ADIPOQ | |
| Mouse myeloma cell line NS0-derived recombinant human Adiponectin/Acrp30 Glu19-Asn244 Accession # Q15848 | |
| 25 μg | |
| Primary | |
| Detects human Adiponectin/Acrp30 in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant mouse Adiponectin is observed and less than 5% cross-reactivity with recombinant rat Adiponectin is observed. | |
| Human | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ACDC, Acrp30, ACRP30ADPN, adipocyte, C1Q and collagen domain containing, adiponectin, adiponectin, C1Q and collagen domain containing, AdipoQ, ADIPQTL1, apM-1, ApM1, APM1APM-1, C1q and collagen domain-containing protein, GBP28, GBP28apM1, Gelatin-binding protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 9370 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido