missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Aggrecan Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems MAB11304-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Aggrecan Monoclonal antibody specifically detects Aggrecan in Human samples. It is validated for ELISA
Especificaciones
| Aggrecan | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| ACAN, AGC1, AGC1SEDK, aggrecan, aggrecan core protein, Cartilage-specific proteoglycan core protein, Chondroitin sulfate proteoglycan 1, Chondroitin sulfate proteoglycan core protein 1, CSPCP, CSPG1, CSPG1aggrecan 1, CSPGCP, large aggregating proteoglycan, MSK16, MSK16AGCAN, SEDK | |
| Mouse myeloma cell line NS0-derived recombinant human Aggrecan, Val20-Gly675, Accession # NP_037359 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG1 |
| ELISA | |
| 179503 | |
| ELISA | |
| NP_037359 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 176 | |
| Human | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido