Learn More
Gibco™ Human BAFF (BLyS) Recombinant Protein
Descripción
Carrier-Free Reconstitution: We recommend that this vial be briefly centrifuged prior to opening to bring the contents to the bottom. Reconstitute in sterile, distilled water to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2°C to 8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein, such as 0.1% BSA and store in working aliquots at -20°C to -80°C. Storage: The lyophilized protein is stable at room temperature for 1 month, but should be kept at -20°C for long term storage. Working aliquots stored with a carrier protein are stable for at least 3 months at -20°C to -80°C. Avoid repeated freeze/thaw cycles.
Especificaciones
Especificaciones
| Número de acceso | Q9Y275 |
| Para utilizar con (aplicación) | Bioactivity |
| Formulación | PBS with 0.5mM DTT and no preservative; pH 7.5 |
| ID de gen (Entrez) | 10673 |
| Peso molecular | 17 kDa |
| Nombre | Human BAFF (BLyS) |
| Intervalo de pH | 7.5 |
| Método de purificación | Purified |
| Cantidad | 20 μg |
| Requisitos de almacenamiento | -20°C |
| Mostrar más |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.