Learn More
Human BAI2 Alexa Fluor 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Rabbit Monoclonal Antibody
Marca:  R&D Systems FAB9338V-100UG
Código de producto. 18753469
  • 491.00€ / 100 microgramos
Descripción
BAI2 Monoclonal specifically detects BAI2 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
BAI2
Monoclonal
Alexa Fluor 405
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
brain-specific angiogenesis inhibitor 2, Brain-specific angiongenesis inhibitor-2
Human embryonic kidney cell, HEK293-derived human BAI2, Phe21-Asp911, Accession # O60241-3
100 μg
Primary
Human
Purified
Flow Cytometry
2528B
Flow Cytometry 0.25-1 ug/10^6 cells
O60241-3
Rabbit
Protein A or G purified from hybridoma culture supernatant
RUO
576
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
IgG
Certificados
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.