missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human BMP-4 Recombinant Protein, PeproTech®

Código de producto. p-7246145
Click to view available options
Cantidad:
10 x 100 μg
100 μg
1 μg
5 x 100 μg
50 μg
5 μg
Tamaño de la unidad:
1 microgramo
1 miligramo
100 microgramos
5 microgramos
50 microgramos
500 microgramos
paquete de 10
paquete de 5
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17800943

Marca: Gibco™ 12005100UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

120-05-1MG will be provided as 10 x 100 μg (120-05-100UG). 120-05-500UG will be provided as 5 x 100 μg (120-05-100UG). Recombinant Human BMP-4, expressed in HeLa cells, is a 25.6 kDa homodimeric glycoprotein. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The protein encoded by this gene is a member of the bone morphogenetic protein family which is part of the transforming growth factor-beta superfamily. The superfamily includes large families of growth and differentiation factors. Bone morphogenetic proteins were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. This particular family member plays an important role in the onset of endochondral bone formation in humans, and a reduction in expression has been associated with a variety of bone diseases, including the heritable disorder Fibrodysplasia Ossificans Progressiva. Alternative splicing in the 5' untranslated region of this gene has been described and three variants are described, all encoding an identical protein.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso P12644
Para utilizar con (aplicación) Control, Western Blot Control, Bioactivity
Formulación Protein with no preservative
ID de gen (Entrez) 652
Peso molecular 25.6 kDa
Nombre Human BMP-4
Método de purificación Purified
Cantidad 100 μg
Fuente HeLa cells
Requisitos de almacenamiento -20°C
Estado normativo RUO
Concentración de endotoxinas <1 EU/μg
Alias de gen BMP; BMP2B; BMP-2B; Bmp2b1; Bmp2b-1; Bmp4; Bmp-4; BOMPR4A; Bone morphogenetic protein; Bone morphogenetic protein 2B; bone morphogenetic protein 4; bone morphogenetic protein 4 preproprotein; DVR4; Dvr-4; MCOPS6; OFC11; ZYME
Nombre común BMP4
Símbolo de gen BMP4
Actividad biológica Determined by its ability to induce alkaline phosphatase production by ATDC-5 cells. The expected ED50 for this effect is 2-8 ng/ml
Tipo de producto Protein
Conjugado Unconjugated
Recombinante Recombinant
Secuencia HHSQRARKKN KNCRRHSLYV DFSDVGWNDW IVAPPGYQAF YCHGDCPFPL ADHLNSTNHA IVQTLVNSVN SSIPKACCVP TELSAISMLY LDEYDKVVLK NYQEMVVEGC GCR
Contenido y almacenamiento -20°C
Actividad Determined by its ability to induce alkaline phosphatase production by ATDC-5 cells. The expected ED50 for this effect is 2-8 ng/ml.
Sistema de expresión HeLa cells
Formulario Lyophilized
Grado de pureza o calidad ≥ 97% by SDS-PAGE gel and HPLC analyses.
Mostrar más Mostrar menos
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado