Learn More
Abnova™ Human BMP3 Partial ORF (NP_001192, 373 a.a. - 472 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00000651-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
BMP3 belongs to the transforming growth factor-beta (TGFB) superfamily. Bone morphogenic protein, also known as osteogenin, induces bone formation. [provided by RefSeq]
Sequence: RYLKVDFADIGWSEWIISPKSFDAYYCSGACQFPMPKSLKPSNHATIQSIVRAVGVVPGIPEPCCVPEKMSSLSILFFDENKNVVLKVYPNMTVESCACREspecificaciones
NP_001192 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RYLKVDFADIGWSEWIISPKSFDAYYCSGACQFPMPKSLKPSNHATIQSIVRAVGVVPGIPEPCCVPEKMSSLSILFFDENKNVVLKVYPNMTVESCACR | |
RUO | |
BMP3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
651 | |
BMP3 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BMP-3A | |
BMP3 | |
Recombinant | |
wheat germ expression system |