Learn More
Invitrogen™ Human BOLL (aa 185-263) Control Fragment Recombinant Protein
Recombinant Protein
Marca: Invitrogen™ RP92306
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This gene belongs to the DAZ gene family required for germ cell development. It encodes an RNA-binding protein which is more similar to Drosophila Boule than to human proteins encoded by genes DAZ (deleted in azoospermia) or DAZL (deleted in azoospermia-like). Loss of this gene function results in the absence of sperm in semen (azoospermia). Histological studies demonstrated that the primary defect is at the meiotic G2/M transition. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.Especificaciones
Q8N9W6 | |
Blocking Assay, Control | |
66037 | |
100 μL | |
RUO | |
BOLL | |
Human | |
ATTQYLPGQWQWSVPQPSASSAPFLYLQPSEVIYQPVEIAQDGGCVPPPLSLMETSVPEPYSDHGVQATYHQVYAPSAI | |
Liquid |
≥5.0 mg/mL | |
1M urea, PBS with no preservative; pH 7.4 | |
BOLL | |
-20° C, Avoid Freeze/Thaw Cycles | |
4930554P13Rik; 4930597B14Rik; bol, boule-like; bol, boule-like (Drosophila); Boll; BOULE; boule homolog, RNA binding protein; boule-like RNA binding protein; boule-like RNA-binding protein; protein boule-like; RGD1559527 | |
Unconjugated | |
Recombinant | |
E. coli |