Learn More
Invitrogen™ Human BOLL (aa 78-177) Control Fragment Recombinant Protein
Recombinant Protein
Marca:  Invitrogen™ RP92305
Código de producto. 30193724
  • 259.00€ / 100 microlitros
Explore más ofertas especiales
Descripción
Descripción
This gene belongs to the DAZ gene family required for germ cell development. It encodes an RNA-binding protein which is more similar to Drosophila Boule than to human proteins encoded by genes DAZ (deleted in azoospermia) or DAZL (deleted in azoospermia-like). Loss of this gene function results in the absence of sperm in semen (azoospermia). Histological studies demonstrated that the primary defect is at the meiotic G2/M transition. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
Q8N9W6
Blocking Assay, Control
66037
100 μL
RUO
BOLL
Human
VTFETQEDAQKILQEAEKLNYKDKKLNIGPAIRKQQVGIPRSSIMPAAGTMYLTTSTGYPYTYHNGVAYFHTPEVTSVPPPWPSRSVCSSPVMVAQPIYQ
Liquid
≥5.0 mg/mL
1M urea, PBS with no preservative; pH 7.4
BOLL
-20° C, Avoid Freeze/Thaw Cycles
4930554P13Rik; 4930597B14Rik; bol, boule-like; bol, boule-like (Drosophila); Boll; BOULE; boule homolog, RNA binding protein; boule-like RNA binding protein; boule-like RNA-binding protein; protein boule-like; RGD1559527
Unconjugated
Recombinant
E. coli