Learn More
Abnova™ Human CAPZA2 Partial ORF (NP_006127, 111 a.a. - 169 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00000830-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene is a member of the F-actin capping protein alpha subunit family. It is the alpha subunit of the barbed-end actin binding protein Cap Z. By capping the barbed end of actin filaments, Cap Z regulates the growth of the actin filaments at the barbed end. [provided by RefSeq]
Sequence: CEVENAVESWRTSVETALRAYVKEHYPNGVCTVYGKKIDGQQTIIACIESHQFQAKNFWEspecificaciones
NP_006127 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.23kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
CEVENAVESWRTSVETALRAYVKEHYPNGVCTVYGKKIDGQQTIIACIESHQFQAKNFW | |
RUO | |
CAPZA2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
830 | |
CAPZA2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CAPPA2/CAPZ | |
CAPZA2 | |
Recombinant | |
wheat germ expression system |