Learn More
ACROBiosystems Human Carbonic Anhydrase II / CA2 Protein, His Tag
302.42€ - 2020.12€
Especificaciones
Número de acceso | NP_000058.1 |
---|---|
Peso molecular | 30.1 kDa |
Fuente | Human CA2, His Tag (CA2-H5228) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Lys 260 (Accession # NP_000058.1). |
Inmunógeno | Met 1 - Lys 260 |
Requisitos de almacenamiento | -20°C |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15876087
|
ACROBiosystems
CA2-H5228-1MG |
1 mg |
2020.12€
1 miligramo |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
15866087
|
ACROBiosystems
CA2-H5228-100UG |
100 μg |
302.42€
100 microgramos |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
Carbonic anhydrases (CAs) are a large family of zinc metalloenzymes. CAs form a family of enzymes that catalyze the rapid interconversion of carbon dioxide and water to bicarbonate and protons (or vice versa), a reversible reaction that occurs rather slowly in the absence of a catalyst. One of the functions of the enzyme in animals is to interconvert carbon dioxide and bicarbonate to maintain acid-base balance in blood and other tissues, and to help transport carbon dioxide out of tissues. The active site of most carbonic anhydrases contains a zinc ion. They are, therefore, classified as metalloenzymes. There are at least five distinct CA families (α, β, γ, δ and ε). These families have no significant amino acid sequence similarity and in most cases are thought to be an example of convergent evolution. The α-CAs are found in humans.Carbonic anhydrase II (CA2) is also known as Carbonate dehydratase II, Carbonic anhydrase C, is one of fourteen forms of human α carbonic anhydrases. Defects in this enzyme are associated with osteopetrosis and renal tubular acidosis. Renal carbonic anhydrase allows the reabsorption of sodium ions in the proximal tubule. Carbonic anhydrase II has been shown to interact with Band 3 and Sodium-hydrogen antiporter 1.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 30.1 kDa. The protein migrates as 31-33 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
NP_000058.1 | |
Human CA2, His Tag (CA2-H5228) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Lys 260 (Accession # NP_000058.1). | |
-20°C | |
Carbonic Anhydrase II | |
Human | |
20 mM Tris, 150 mM NaCl, pH8.0 | |
HEK293 | |
Bio-Markers and CD Antigens |
30.1 kDa | |
Met 1 - Lys 260 | |
CA2,CA-II,CAII,Car2 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |