Learn More
ACROBiosystems Human Carbonic Anhydrase IX / CA9 (38-414) Protein, His Tag (active enzyme)
423.38€ - 2951.55€
Especificaciones
Número de acceso | NP_001207.2 |
---|---|
Peso molecular | 41.7 kDa |
Fuente | Human Carbonic Anhydrase IX (38-414), His Tag (CA9-H5226) is expressed from human 293 cells (HEK293). It contains AA Gln 38 - Asp 414 (Accession # Q16790-1). |
Inmunógeno | Gln 38 - Asp 414 |
Requisitos de almacenamiento | -20°C |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15876097
|
ACROBiosystems
CA9-H5226-1MG |
1 mg |
2951.55€
1 miligramo |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
15886097
|
ACROBiosystems
CA9-H5226-50UG |
50 μg |
423.38€
50 microgramos |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
Carbonic anhydrases (CAs) are a large family of zinc metalloenzymes. CAs form a family of enzymes that catalyze the rapid interconversion of carbon dioxide and water to bicarbonate and protons (or vice versa), a reversible reaction that occurs rather slowly in the absence of a catalyst. One of the functions of the enzyme in animals is to interconvert carbon dioxide and bicarbonate to maintain acid-base balance in blood and other tissues, and to help transport carbon dioxide out of tissues. The active site of most carbonic anhydrases contains a zinc ion. There are at least five distinct CA families (α, β, γ, δ and ε). Carbonic anhydrase 9 (CA9 / CAIX) is also known as Membrane antigen MN (MN), Renal cell carcinoma-associated antigen G250, which belongs to the alpha-carbonic anhydrase family. CA9 / CAIX with an optimal activity at pH 6.49. Reversible hydration of carbon dioxide. CA IX participates in pH regulation. CA9 may be involved in the control of cell proliferation and transformation. CA-IX appears to be a novel specific biomarker for a cervical neoplasia.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 41.7 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
NP_001207.2 | |
Human Carbonic Anhydrase IX (38-414), His Tag (CA9-H5226) is expressed from human 293 cells (HEK293). It contains AA Gln 38 - Asp 414 (Accession # Q16790-1). | |
-20°C | |
Carbonic Anhydrase IX | |
Human | |
20 mM MES, 100 mM NaCl, pH6.5 | |
HEK293 | |
Bio-Markers and CD Antigens |
41.7 kDa | |
Gln 38 - Asp 414 | |
CAIX,CA9,CA-IX,G250,MN,P54/58 N,pMW1 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |