missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human CCL21 (6Ckine) Recombinant Protein, PeproTech®
Click to view available options
Cantidad:
100 μg
1 mg
20 μg
250 μg
500 μg
50 μg
5 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
20 microgramos
250 microgramos
5 microgramos
50 microgramos
500 microgramos
Descripción
Recombinant Human Exodus-2 (CCL21) is a 12.2 kDa protein containing 111 amino acid residues. *Human Exodus-2 (Catalog Number 300-35A) has replaced Human Exodus-2 (Catalog Number 300-35). This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
This gene is one of several CC cytokine genes clustered on the p-arm of chromosome 9. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. Similar to other chemokines the protein encoded by this gene inhibits hemopoiesis and stimulates chemotaxis. This protein is chemotactic in vitro for thymocytes and activated T cells, but not for B cells, macrophages, or neutrophils. The cytokine encoded by this gene may also play a role in mediating homing of lymphocytes to secondary lymphoid organs. It is a high affinity functional ligand for chemokine receptor 7 (CCR7) that is expressed on T and B lymphocytes and a known receptor for another member of the cytokine family (small inducible cytokine A19).
Especificaciones
Especificaciones
| Número de acceso | O00585 |
| Para utilizar con (aplicación) | Functional Assay |
| Formulación | protein with no preservative |
| ID de gen (Entrez) | 6366 |
| Peso molecular | 12.2 kDa |
| Nombre | Human CCL21 (6 Ckine) |
| Cantidad | 100 μg |
| Fuente | E. coli |
| Estado normativo | RUO |
| Concentración de endotoxinas | <1 EU/ μg |
| Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido