Learn More
Invitrogen™ Human CEA ProQuantum Immunoassay Kit
The ProQuantum Human CEA Immunoassay Kit is designed to provide quantitative measurements of human CEA in small sample volumes.
Marca: Invitrogen™ A46341
Detalles adicionales : Peso : 0.50000kg
Descripción
Utilizing proximity-based amplification technology, the assay combines the analyte specificity of high-affinity antibody-antigen binding with the signal detection and amplification capabilities of real-time PCR to achieve a simple yet powerful next-generation protein quantitation platform. A user-friendly workflow combined with intuitive software for analytics enables sample-to-answer in just 2 hours.
- High sensitivity-detect low levels of protein with greater sensitivity than traditional methods
- Broad dynamic range-5 logarithmic units, minimizing sample dilutions to ensure they fall within the range
- Small sample consumption-use 2–5 μL of sample (compared to 75 μL for triplicate wells with other methods)
- Fast, easy workflow-2 hours from sample to answer, with no wash steps
- No proprietary instrument to purchase-runs on any real-time PCR instrument
Especificaciones
1600029H12Rik; antígeno carcinoembrionario; molécula 5 de adhesión celular relacionada con antígeno carcinoembrionario; molécula 5 de adhesión celular relacionada con antígeno carcinoembrionario; CD66e; CEA; CEACAM5; antígeno meconio 100; OTTHUMP00000199033; glicoproteína 30 específica para el embarazo; Psg30; sCD66e; CD66e soluble | |
<0,3 pg/ml | |
Instrumentos qPCR | |
Ceacam5 | |
96 pruebas | |
Suero, 2-5 µl; plasma, 2-5 µl; sobrenadante, 2-5 µl | |
CD66e (CEACAM5) | |
2 h |
De 0,32 a 25 000 pg/ml | |
ELISA | |
P06731 | |
Tampón de dilución de ensayo Conjugado de anticuerpos A Conjugado de anticuerpos B Tampón de dilución de conjugado de anticuerpos Estándar Mezcla maestra Ligasa | |
Para uso exclusivo en investigación | |
-20 °C | |
30 min |