missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CELSR3 Partial ORF (NP_001398, 71 a.a. - 180 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

335.00€ - 508.00€

Especificaciones

Número de acceso NP_001398
Para utilizar con (aplicación) Antibody Production, ELISA, Protein Array, Western Blot
Formulación 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
ID de gen (Entrez) 1951
Peso molecular 37.84kDa
Ver más especificaciones

Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
16174401
Ver documentos Detalles de la promoción
Abnova™
H00001951-Q01.25UG
25 ug
508.00€
25 microgramos
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 04-09-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
16164401
Ver documentos Detalles de la promoción
Abnova™
H00001951-Q01.10UG
10 ug
335.00€
10 microgramos
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 04-09-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Descripción

Descripción

The protein encoded by this gene is a member of the flamingo subfamily, part of the cadherin superfamily. The flamingo subfamily consists of nonclassic-type cadherins; a subpopulation that does not interact with catenins. The flamingo cadherins are located at the plasma membrane and have nine cadherin domains, seven epidermal growth factor-like repeats and two laminin A G-type repeats in their ectodomain. They also have seven transmembrane domains, a characteristic unique to this subfamily. It is postulated that these proteins are receptors involved in contact-mediated communication, with cadherin domains acting as homophilic binding regions and the EGF-like domains involved in cell adhesion and receptor-ligand interactions. The specific function of this particular member has not been determined. [provided by RefSeq]

Sequence: REDGGPGLGVREPIFVGLRGRRQSARNSRGPPEQPNEELGIEHGVQPLGSRERETGQGPGSVLYWRPEVSSCGRTGPLQRGSLSPGALSSGVPGSGNSSPLPSDFLIRHH
Especificaciones

Especificaciones

NP_001398
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CDHF11/EGFL1/FMI1/HFMI1/MEGF2/RESDA1
CELSR3
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
1951
CELSR3 (Human) Recombinant Protein (Q01)
REDGGPGLGVREPIFVGLRGRRQSARNSRGPPEQPNEELGIEHGVQPLGSRERETGQGPGSVLYWRPEVSSCGRTGPLQRGSLSPGALSSGVPGSGNSSPLPSDFLIRHH
RUO
CELSR3
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documentos

Documentos

Product Certifications