Learn More
ACROBiosystems Human CG alpha / CGA Protein, His Tag
423.38€ - 2661.24€
Especificaciones
Número de acceso | AAH20782 |
---|---|
Peso molecular | 12.1 kDa |
Fuente | Human CG alpha, His Tag (CGA-H52H2) is expressed from human 293 cells (HEK293). It contains AA Ala 25 - Ser 116 (Accession # AAH20782). |
Inmunógeno | Ala 25 - Ser 116 |
Requisitos de almacenamiento | -20°C |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
16477419
|
ACROBiosystems
CGA-H52H2-1MG |
1 mg |
2661.24€
1 miligramo |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
15876437
|
ACROBiosystems
CGA-H52H2-100UG |
100 μg |
423.38€
100 microgramos |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
Chorionic gonadotrophin subunit alpha (CGA),a member of the glycoprotein hormones subunit alpha family, is also known as glycoprotein hormones alpha chain, choriogonadotropin alpha chain, follicle-stimulating hormone alpha chain (FSH-alpha), luteinizing hormone alpha chain (LSH-alpha) and thyroid-stimulating hormone alpha chain (TSH-alpha). The hormones human chorionic gonadotropin (hCG), luteinizing hormone (LH), follicle-stimulating hormone (FSH), and thyroid-stimulating hormone (TSH) are dimers consisting of alpha and beta subunits that are associated noncovalently. The alpha subunits (CGA) of these four human glycoprotein hormones are nearly identical, however, their beta chains are unique and confer biological specificity.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 12.1 kDa. The protein migrates as 22-26 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
AAH20782 | |
Human CG alpha, His Tag (CGA-H52H2) is expressed from human 293 cells (HEK293). It contains AA Ala 25 - Ser 116 (Accession # AAH20782). | |
-20°C | |
CG alpha | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Cytokines and Growth Factors |
12.1 kDa | |
Ala 25 - Ser 116 | |
CGA,TSH-alpha,LSH-alpha,CG-alpha,FSH-alpha | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>92% as determined by SDS-PAGE |