missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Collagen IV alpha 1 Antibody, R&D Systems™
Sheep Polyclonal Antibody has been used in 1 publication
Marca: R&D Systems AF6308
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Collagen IV alpha 1 Polyclonal specifically detects Collagen IV alpha 1 in Human samples. It is validated for Western Blot.Especificaciones
Collagen IV alpha 1 | |
Polyclonal | |
Unconjugated | |
P02462 | |
COL4A1 | |
Mouse myeloma cell line NS0-derived recombinant human Collagen IV alpha 1 Ser1441-Thr1669 Accession # P02462 | |
100 μg | |
Primary | |
Detects human Collagen IV alpha 1 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse COL13A1, recombinant human (rh) COL25A1 and rhCOL23A1 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
alpha-1 chain, arresten, COL4A1, COL4A1 NC1 domain, collagen alpha-1(IV) chain, collagen IV, alpha-1 polypeptide, collagen, type IV, alpha 1, Collagen-4, EC 3.4.23, EC 6.3.1.2 | |
Sheep | |
Affinity Purified | |
RUO | |
1282 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido