Learn More
Novus Biologicals™ Human Colon Tissue MicroArray (Tumor)
Tissue arrays are 4 - 6 um in thickness, and mounted on positively charged glass slides
Marca: Novus Biologicals™ NBP2-42062
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
5 Slides | |
Human | |
Immunohistochemistry, Immunohistochemistry-Paraffin |