missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CPZ (aa 387-461) Control Fragment Recombinant Protein
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Descripción
Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-140217 (PA5-140217. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
CPZ is a member of the metallocarboxypeptidase family. This enzyme displays carboxypeptidase activity towards substrates with basic C-terminal residues. It is most active at neutral pH and is inhibited by active site-directed inhibitors of metallocarboxypeptidases.
Especificaciones
Especificaciones
| Número de acceso | Q66K79 |
| Concentración | ≥5.0 mg/mL |
| Para utilizar con (aplicación) | Blocking Assay, Control |
| Formulación | 1 M urea, PBS with no preservative; pH 7.4 |
| ID de gen (Entrez) | 8532 |
| Nombre | Human CPZ (aa 387-461) Control Fragment |
| Cantidad | 100 μL |
| Estado normativo | RUO |
| Alias de gen | AV136066; carboxypeptidase Z; Cpz; metallocarboxypeptidase Z |
| Nombre común | CPZ |
| Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu