missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CX3CL1 Synthetic Peptide
Synthetic Peptide
Marca: Invitrogen™ PEP0398
Detalles adicionales : Peso : 0.01000kg
Descripción
CX3CL1 Synthetic Peptide, PEP-0398, from Invitrogen.
This peptide corresponds to 18 amino acids near the amino terminus of human CX3CL1.PEP-0398 can be used as a blocking peptide with polyclonal antibody PA5-20296.Especificaciones
6376 | |
purified | |
Human CX3CL1 | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
P78423 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
CX3CL1 | |
0.02% sodium azide | |
50 μg | |
PBS |