Learn More
ACROBiosystems Human Decorin Protein, His Tag
338.70€ - 1463.68€
Especificaciones
Número de acceso | NP_001911.1 |
---|---|
Peso molecular | 38.8 kDa |
Fuente | Human Decorin, His Tag (DE1-H5223) is expressed from human 293 cells (HEK293). It contains AA Gly 17 - Lys 359 (Accession # NP_001911.1). |
Inmunógeno | Gly 17 - Lys 359 |
Requisitos de almacenamiento | -20°C |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15886557
|
ACROBiosystems
DE1-H5223-200UG |
200 μg |
338.70€
200 microgramos |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
15876557
|
ACROBiosystems
DE1-H5223-1MG |
1 mg |
1463.68€
1 miligramo |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Description
Decorin is also known as bone proteoglycan II, PGS2, SLRR1B, DCN, DSPG2 and PG40, is a secreted chondroitin /dermatan sulfate proteoglycan in the family of small leucine-rich proteoglycans (SLRPs). Decorin is a small cellular or pericellular matrix proteoglycan and is closely related in structure to biglycan protein. Decorin and biglycan are thought to be the result of a gene duplication. This protein is a component of connective tissue, binds to type I collagen fibrils, and plays a role in matrix assembly. Decorin appears to influence fibrillogenesis, and also interacts with fibronectin, thrombospondin, the complement component C1q, epidermal growth factor receptor (EGFR) and transforming growth factor-beta (TGF-beta). Defects in DCN are the cause of congenital stromal corneal dystrophy (CSCD).
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 38.8 kDa. The protein migrates as 46-55 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.Specifications
NP_001911.1 | |
Human Decorin, His Tag (DE1-H5223) is expressed from human 293 cells (HEK293). It contains AA Gly 17 - Lys 359 (Accession # NP_001911.1). | |
-20°C | |
Decorin | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Other Recombinant Proteins |
38.8 kDa | |
Gly 17 - Lys 359 | |
Decorin,DCN,CSCD,DSPG2,PG40,PGII,PGS2,SLRR1B | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |