missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human E-selectin (CD62E) Recombinant Protein, PeproTech®

Código de producto. 17861633
Click to view available options
Cantidad:
100 μg
10 μg
1 mg
250 μg
500 μg
50 μg
Tamaño de la unidad:
1 miligramo
10 microgramos
100 microgramos
250 microgramos
50 microgramos
500 microgramos
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17861633

Marca: Gibco™ 15015250UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

Recombinant Human E-selectin is a 58.6 kDa protein containing 535 amino acid residues, corresponding to the extracellular portion of the full length protein. Due to glycosylation, E-selectin migrates at an apparent molecular weight of approximately 65-85 kDa by SDS-PAGE analysis under reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Leukocytes adhere to the blood vessel endothelium during extravasation in postcapillary venules of lymph nodes. In addition, leukocyte adhesion occurs in the capillaries and small venules at any site in the body after onset of inflammation. This response is immediate and involves a cascade of adhesion receptors. At the endothelial surface members of the selectin and immunoglobulin superfamilies participate in this cascade. Selectins are C-type cell surface lectins that play a role in leukocyte adhesion to the blood vessel wall endothelium. E-selectin (CD62E) is an endothelial cell specific selectin that is expressed only after activation with proinflammatory cytokines. In vitro experiments have shown that IL-1, TNFalpha and bacterial wall components like lipopolysaccharides induce the transcription of E-selectin in a NFkB dependent signaling cascade. E-selectin has been associated with blood vessel endothelium in diverse inflammatory situations.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso P16581
Para utilizar con (aplicación) Functional Assay
Formulación protein with no preservative
ID de gen (Entrez) 6401
Peso molecular 58.6 kDa
Nombre Human E-selectin (CD62E)
Cantidad 250 μg
Fuente CHO cells
Estado normativo RUO
Concentración de endotoxinas <1 EU/ μg
Alias de gen CD62 antigen-like family member E; CD62E; e selectin; Elam; ELAM1; ELAM-1; endothelial adhesion molecule 1; endothelial leukocyte adhesion molecule; endothelial leukocyte adhesion molecule 1; endothelial leukocyte adhesion molecule I; ESEL; E-selectin; LECAM2; leukocyte endothelial cell adhesion molecule 2; leukocyte-endothelial cell adhesion molecule 2; RP1-117P20.2; SELE; selectin E; selectin E (endothelial adhesion molecule 1); selectin, endothelial cell
Nombre común E-selectin (CD62E)
Símbolo de gen SELE
Actividad biológica Measured by its ability to support adhesion of U937 cells, a human hystiocytic lymphoma cell line.
Conjugado Unconjugated
Recombinante Recombinant
Secuencia WSYNTSTEAM TYDEASAYCQ QRYTHLVAIQ NKEEIEYLNS ILSYSPSYYW IGIRKVNNVW VWVGTQKPLT EEAKNWAPGE PNNRQKDEDC VEIYIKREKD VGMWNDERCS KKKLALCYTA ACTNTSCSGH GECVETINNY TCKCDPGFSG LKCEQIVNCT ALESPEHGSL VCSHPLGNFS YNSSCSISCD RGYLPSSMET MQCMSSGEWS APIPACNVVE CDAVTNPANG FVECFQNPGS FPWNTTCTFD CEEGFELMGA QSLQCTSSGN WDNEKPTCKA VTCRAVRQPQ NGSVRCSHSP AGEFTFKSSC NFTCEEGFML QGPAQVECTT QGQWTQQIPV CEAFQCTALS NPERGYMNCL PSASGSFRYG SSCEFSCEQG FVLKGSKRLQ CGPTGEWDNE KPTCEAVRCD AVHQPPKGLV RCAHSPIGEF TYKSSCAFSC EEGFELHGST QLECTSQGQW TEEVPSCQVV KCSSLAVPGK INMSCSGEPV FGTVCKFACP EGWTLNGSAA RTCGATGHWS GLLPTCEAPT ESNIP
Contenido y almacenamiento -20°C
Sistema de expresión CHO cells
Formulario Lyophilized
Grado de pureza o calidad ≥ 95% by SDS-PAGE gel and HPLC analyses.
Mostrar más Mostrar menos
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado