missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human Ephrin-A1 / EFNA1 Protein, Fc Tag
302.42€ - 2020.12€
Especificaciones
Número de acceso | NP_004419.2 |
---|---|
Peso molecular | 46.0 kDa |
Fuente | Human Ephrin-A1, Fc Tag (EF1-H5251) is expressed from human 293 cells (HEK293). It contains AA Asp 19 - Ser 182 (Accession # P20827-1). |
Inmunógeno | Asp 19 - Ser 182 |
Requisitos de almacenamiento | -20°C |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15896607
|
ACROBiosystems
EF1-H5251-1MG |
1 mg |
2020.12€
1 miligramo |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
15886607
|
ACROBiosystems
EF1-H5251-100UG |
100 μg |
302.42€
100 microgramos |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
Ephrin-A1 (EFNA1) is also known as EPLG1, LERK1, TNFAIP4, Immediate early response protein B61, which belongs to the ephrin family. EFNA1 contains one ephrin RBD (ephrin receptor-binding) domain. EFNA1 is cell surface GPI-bound ligand for Eph receptors, and is a family of receptor tyrosine kinases which are crucial for migration, repulsion and adhesion during neuronal, vascular and epithelial development. EFNA1 binds promiscuously Eph receptors residing on adjacent cells, leading to contact-dependent bidirectional signaling into neighboring cells. EFNA1 plays an important role in angiogenesis and tumor neovascularization.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 46.0 kDa. As a result of glycosylation, the protein migrates as 53-57 kDa under reducing (R) condition, and 100-115 kDa under non-reducing (NR) condition (SDS-PEspecificaciones
NP_004419.2 | |
Human Ephrin-A1, Fc Tag (EF1-H5251) is expressed from human 293 cells (HEK293). It contains AA Asp 19 - Ser 182 (Accession # P20827-1). | |
-20°C | |
Ephrin-A1 | |
Human | |
50 mM Tris, 100 mM Glycine, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |
46.0 kDa | |
Asp 19 - Ser 182 | |
EFNA1,Ephrin-A1,LERK-1,TNFAIP4,EPLG1,B61,ECKLG,EFL1 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |