Learn More
Gibco™ Human EPO (Erythropoietin) Recombinant Protein, PeproTech®
CHO cells Recombinant Protein
Marca: Gibco™ 100-64-500UG
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Erythropoietin (EPO) protein is a growth factor essential for the survival, proliferation, and differentiation of erythroid precursors. EPO is not responsible for early commitment to the lineage, and instead induces the final maturation of partially differentiated erythroid cells and the production of red cell-specific proteins, such as hemoglobin. The EPO receptor (EPOR) is present in higher levels on precursor cells, and expression diminishes with cell maturation.Hepatocytes are the main source of EPO in the fetus, followed by interstitial cells of the kidney after birth and through adulthood. EPO expression is upregulated during hypoxia, and low levels of transcription have also been detected in lung and brain tissue under these conditions. Recent data suggest that EPO may also play a role in the protection of neuronal tissue from damage and initiation of repair.Especificaciones
P01588 | |
protein with no preservative | |
18.4 | |
500 μg | |
-20°C | |
<1 EU/μg | |
EPO | |
Unconjugated | |
APPRLICDSR VLERYLLEAK EAENITTGCA EHCSLNENIT VPDTKVNFYA WKRMEVGQQA VEVWQGLALL SEAVLRGQAL LVNSSQPWEP LQLHVDKAVS GLRSLTTLLR ALGAQKEAIS PPDAASAAPL RTITADTFRK LFRVYSNFLR GKLKLYTGEA CRTGDR | |
≥ 90% by SDS-PAGE gel and HPLC analyses. |
ELISA, Functional Assay, Western Blot | |
2056 | |
Human EPO (Erythropoietin) | |
CHO cells | |
RUO | |
EP; EPO; Epoetin; erythropoietin; MGC138142; MVCD2 | |
Human | |
Recombinant | |
Lyophilized |