missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human FBXL16 Full-length ORF (NP_699181.1, 1 a.a. - 479 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00146330-P01L
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Members of the F-box protein family, such as FBXL16, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).[supplied by OMIM]
Sequence: MSSPGIDGDPKPPCLPRNGLVKLPGQPNGLGAASITKGTPATKNRPCQPPPPPTLPPPSLAAPLSRAALAGGPCTPAGGPASALAPGHPAERPPLATDEKILNGLFWYFSACEKCVLAQVCKAWRRVLYQPKFWAGLTPVLHAKELYNVLPGGEKEFVNLQGFAARGFEGFCLVGVSDLDICEFIDNYALSKKGVKAMSLKRSTITDAGLEVMLEQMQGVVRLELSGCNDFTEAGLWSSLSARITSLSVSDCINVADDAIAAISQLLPNLAELSLQAYHVTDTALAYFTARQGHSTHTLRLLSCWEITNHGVVNVVHSLPNLTALSLSGCSKVTDDGVELVAENLRKLRSLDLSWCPRITDMALEYVACDLHRLEELVLDRCVRITDTGLSYLSTMSSLRSLYLRWCCQVQDFGLKHLLALGSLRLLSPAGCPLLTTTGLSGLVQLQELEELELTNCPGATPELFKYFSQHLPRCLVIEEspecificaciones
NP_699181.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
78kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C16orf22/FLJ33735/Fbl16/MGC33974/c380A1.1 | |
FBXL16 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
146330 | |
FBXL16 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSSPGIDGDPKPPCLPRNGLVKLPGQPNGLGAASITKGTPATKNRPCQPPPPPTLPPPSLAAPLSRAALAGGPCTPAGGPASALAPGHPAERPPLATDEKILNGLFWYFSACEKCVLAQVCKAWRRVLYQPKFWAGLTPVLHAKELYNVLPGGEKEFVNLQGFAARGFEGFCLVGVSDLDICEFIDNYALSKKGVKAMSLKRSTITDAGLEVMLEQMQGVVRLELSGCNDFTEAGLWSSLSARITSLSVSDCINVADDAIAAISQLLPNLAELSLQAYHVTDTALAYFTARQGHSTHTLRLLSCWEITNHGVVNVVHSLPNLTALSLSGCSKVTDDGVELVAENLRKLRSLDLSWCPRITDMALEYVACDLHRLEELVLDRCVRITDTGLSYLSTMSSLRSLYLRWCCQVQDFGLKHLLALGSLRLLSPAGCPLLTTTGLSGLVQLQELEELELTNCPGATPELFKYFSQHLPRCLVIE | |
RUO | |
FBXL16 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |