Learn More
ACROBiosystems Human FGF-9 Protein, Fc Tag
266.13€ - 2201.57€
Especificaciones
Número de acceso | AAI03979 |
---|---|
Peso molecular | 50.1 kDa |
Fuente | Human FGF-9, Fc Tag (FG9-H4267) is expressed from human 293 cells (HEK293). It contains AA Leu 4 - Ser 208 (Accession # AAI03979). |
Inmunógeno | Leu 4 - Ser 208 |
Requisitos de almacenamiento | -20°C |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15896757
|
ACROBiosystems
FG9-H4267-100UG |
100 μg |
266.13€
100 microgramos |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
15806767
|
ACROBiosystems
FG9-H4267-1MG |
1 mg |
2201.57€
1 miligramo |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
Fibroblast growth factor 9 is also known as FGF9, GAF, HBFG-9, SYNS3, and is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. FGF9 is also a mitogen for oligodendrocyte type 2 astrocyte progenitor cells, smooth muscle cells, pheochromocytoma PC12 cells, and BALB/3T3 fibroblasts. However, unlike FGF acidic and basic, FGF9 has no effect on human umbilical vein endothelial cells, and it has been demonstrated that FGF9 binds preferentially to the IIIc form of FGFR3. Although no typical signal sequence was found in FGF9, it is secreted efficiently after synthesis not in a conventional manner. In nervous system, FGF9 is produced mainly by neurons and may plays an important role in CNS development. FGF9 has been shown to interact with Fibroblast growth factor receptor 3.
This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 50.1 kDa. As a result of glycosylation, the protein migrates as 55 kDa under reducing (R) condition, and 110 kDa under non-reducing (NR) condition (SDS-PAGE).Especificaciones
AAI03979 | |
Human FGF-9, Fc Tag (FG9-H4267) is expressed from human 293 cells (HEK293). It contains AA Leu 4 - Ser 208 (Accession # AAI03979). | |
-20°C | |
FGF-9 | |
Human | |
50 mM Tris, 100 mM Glycine, pH7.5 | |
HEK293 | |
Cytokines and Growth Factors |
50.1 kDa | |
Leu 4 - Ser 208 | |
FGF9,GAF,HBFG-9,MGC119914,MGC119915,SYNS3 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>92% as determined by SDS-PAGE |