Learn More
ACROBiosystems Human FOLR1 Protein, Strep Tag
Marca: ACROBiosystems FO1-H528B-250UG
Detalles adicionales : Peso : 0.00001kg
Descripción
Folate Receptor 1 (FOLR1) is also known as Folate receptor alpha, Folate Binding Protein (FBP), FOLR, and is a member of the folate receptor (FOLR) family. Members of this gene family have a high affinity for folic acid and for several reduced folic acid derivatives, and mediate delivery of 5-methyltetrahydrofolate to the interior of cells. Mature FOLR1 is an N-glycosylated protein that is anchored to the cell surface by a GPI linkage. FOLR1 is predominantly expressed on epithelial cells and is dramatically upregulated on many carcinomas. FOLR1 is internalized to the endosomal system where it dissociates from its ligand before recycling to the cell surface. A soluble form of FOLR1 can be proteolytically shed from the cell surface into the serum and breast milk. Defects in FOLR1 are the cause of neurodegeneration due to cerebral folate transport deficiency (NCFTD). NCFTD is an autosomal recessive disorder resulting from brain-specific folate deficiency early in life.
This protein carries a twin strep tag at the C-terminus. The protein has a calculated MW of 27.6 kDa. The protein migrates as 33-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
NP_057937.1 | |
250 μg | |
Arg 25 - Met 233 | |
FOLR-1,FBP,FOLR | |
Unconjugated | |
Strep Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
27.6 kDa | |
Human FOLR1, Strep Tag (FO1-H528b) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Met 233 (Accession # NP_057937.1). | |
-20°C | |
FOLR1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |