missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human Frizzled-5 / FZD5 Protein, Fc Tag
314.52€ - 2020.12€
Especificaciones
Número de acceso | Q13467 |
---|---|
Peso molecular | 42.6 kDa |
Fuente | Human Frizzled-5, Fc Tag (FZ5-H5259) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Pro 167 (Accession # Q13467). |
Inmunógeno | Ala 27 - Pro 167 |
Requisitos de almacenamiento | -20°C |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15886797
|
ACROBiosystems
FZ5-H5259-1MG |
1 mg |
2020.12€
1 miligramo |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
15876797
|
ACROBiosystems
FZ5-H5259-100UG |
100 μg |
314.52€
100 microgramos |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
Frizzled-5 (FZD5) is also known as FzE5, which belongs to the G-protein coupled receptor Fz/Smo family. Most of frizzled receptors are coupled to the beta-catenin canonical signaling pathway, which leads to the activation of disheveled proteins, inhibition of GSK-3 kinase, nuclear accumulation of beta-catenin and activation of Wnt target genes. FZD5 contains one FZ (frizzled) domain. FZD5 may be involved in transduction and intercellular transmission of polarity information during tissue morphogenesis and/or in differentiated tissues. FZD5 interacts specifically with Wnt5A to induce the beta-catenin pathway. FZD5 interacts with GOPC.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 42.6 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
Q13467 | |
Human Frizzled-5, Fc Tag (FZ5-H5259) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Pro 167 (Accession # Q13467). | |
-20°C | |
Frizzled-5 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |
42.6 kDa | |
Ala 27 - Pro 167 | |
FZD5,Frizzled-5,C2orf31,,Fz-5,hFz5,FzE5 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |