missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Glut2 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
Marca: R&D Systems MAB1414-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Glut2 Monoclonal specifically detects Glut2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.
Especificaciones
| Glut2 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, CyTOF-ready | |
| P11168 | |
| SLC2A2 | |
| NS0 mouse myeloma cell line transfected with human Glut2 Met1-Val524 Accession # P11168 | |
| 25 μg | |
| Primary | |
| Recognizes human Glut2 expression on human Glut2-transfected NS0 cells, but not on control transfectants. Was shown to detect Glut2 on the surface of the human intestinal cell line Caco2 (2) in flow cytometry and on fixed cells in immunocytochemistry tests. Based on flow cytometric tests on transfected cells, this antibody has no cross-reactivity with human Glut1 or human Glut3. | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 199017 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| GLUT-2, GLUT2 Glucose transporter type 2, liver, SLC2A2, solute carrier family 2 (facilitated glucose transporter), member 2, solute carrier family 2, facilitated glucose transporter member 2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6514 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido