missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human HGF R / c-MET Protein, Fc Tag
MALS verified
459.67€ - 1935.44€
Especificaciones
Número de acceso | AAI30421.1 |
---|---|
Peso molecular | 127.8 kDa |
Fuente | Human HGF R, Fc Tag (MET-H5256) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Thr 932 (Accession # AAI30421.1). |
Inmunógeno | Glu 25 - Thr 932 |
Requisitos de almacenamiento | -20°C |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15887297
|
ACROBiosystems
MET-H5256-1MG |
1 mg |
1935.44€
1 miligramo |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
15897297
|
ACROBiosystems
MET-H5256-200UG |
200 μg |
459.67€
200 microgramos |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
Hepatocyte growth factor receptor (HGFR) is also known as mesenchymal-epithelial transition factor (MET), c-Met, and is a glycosylated receptor tyrosine kinase that plays a central role in epithelial morphogenesis and cancer development. HGFR protein possesses tyrosine-kinase activity. The primary single chain precursor protein is post-translationally cleaved to produce the alpha and beta subunits, which are disulfide linked to form the mature receptor. HGFR is normally expressed by cells of epithelial origin, while expression of HGF is restricted to cells of mesenchymal origin. Upon HGF stimulation, HGFR induces several biological responses that collectively give rise to a program known as invasive growth. Abnormal HGFR activation in cancer correlates with poor prognosis, where aberrantly active HGFR triggers tumor growth, formation of new blood vessels (angiogenesis) that supply the tumor with nutrients, and cancer spread to other organs (metastasis). HGFR is deregulated in many types of human malignancies, including cancers of kidney, liver, stomach, breast, and brain. Normally, only stem cells and progenitor cells express HGFR, However, cancer stem cells are thought to hijack the ability of normal stem cells to express HGFR, and thus become the cause of cancer persistence and spread to other sites in the body. Various mutations in the HGFR gene are associated with papillary renal carcinoma. HGFR mediates a complex program known as invasive growth. Activation of HGFR triggers mitogenesis, and morphogenesis. Human HGF R, Fc Tag is fused with Fc fragment of human IgG1 at the C-terminus.The mature form of HGF R is a disulfide-linked heterodimer composed of proteolytically cleaved α and β chain. Each α and β chain has a calculated MW of 32.5 kDa (α chain) and 9Especificaciones
AAI30421.1 | |
Human HGF R, Fc Tag (MET-H5256) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Thr 932 (Accession # AAI30421.1). | |
-20°C | |
HGF R | |
Human | |
50 mM Tris, 100 mM Glycine, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |
127.8 kDa | |
Glu 25 - Thr 932 | |
MET,AUTS9,HGFR,RCCP2,c-Met | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |