Learn More
STEMCELL Technologies Human IgG ELISA Antibody Pair Kit
For detection and measurement of human immunoglobulin G
Marca: STEMCELL Technologies 01994
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- The Human Immunoglobulin G (IgG) ELISA Antibody Pair Kit is intended for those who want the flexibility of setting up their own ELISA assay
- This kit includes capture and detection antibodies and an IgG standard
- It is designed for the quantitative detection and measurement of total human IgG in biological fluids such as serum, plasma, and cell culture supernatants
- IgG is the major immunoglobulin found in blood, lymph fluid, cerebrospinal fluid, and peritoneal fluid
- IgG plays a key role in the humoral immune response, plays a critical step in the opsonization process by interacting with the Fcγ receptor on macrophages, neutrophils, and natural killer cells, and is able to activate the complement system
- NOTE: This kit includes sufficient reagents for 6 x 96-well ELISA plates (Catalog #38019)
- ELISA plates and pNPP Substrate (Catalog #01917) are required for use with the Human IgG ELISA Antibody Pair Kit and are available for purchase separately
Especificaciones
B Cells; Hybridomas | |
1 Kit for 6 Plates |