missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human IL-18BP Protein, Fc Tag
350.80€ - 1318.52€
Especificaciones
Número de acceso | NP_001138529 |
---|---|
Peso molecular | 44.3 kDa |
Fuente | Human IL-18 BP, Fc Tag (ILP-H5253) is expressed from human 293 cells (HEK293). It contains AA Thr 31 - Gly 194 (Accession # NP_001138529). |
Inmunógeno | Thr 31 - Gly 194 |
Requisitos de almacenamiento | -20°C |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15877107
|
ACROBiosystems
ILP-H5253-200UG |
200 μg |
350.80€
200 microgramos |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
15867107
|
ACROBiosystems
ILP-H5253-1MG |
1 mg |
1318.52€
1 miligramo |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
Interleukin-18-binding protein (IL18BP) is also known as Tadekinig-alfa, IL18BPa, which functions as an IL18 inhibitor. IL18BP is a secreted protein which contains one Ig-like C2-type (immunoglobulin-like) domain. IL18BP does not have a transmembrane domain and hence is not anchored to the cell membrane. IL18BP is strongly expressed in heart, lung, placenta and spleen. Isoform A of IL18BP binds to IL-18 and inhibits its activity. IL18BP also functions as an inhibitor of the early TH1 cytokine response. IL18BP suppresses the production of IFN-gamma resulting in reduced T-helper type 1 immune responses.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 44.3 kDa. The protein migrates as 56-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
NP_001138529 | |
Human IL-18 BP, Fc Tag (ILP-H5253) is expressed from human 293 cells (HEK293). It contains AA Thr 31 - Gly 194 (Accession # NP_001138529). | |
-20°C | |
IL-18 BP | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |
44.3 kDa | |
Thr 31 - Gly 194 | |
IL18BP,IL18BPa,Tadekinig-alfa | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |