Learn More
ACROBiosystems Human IL-2 R alpha / CD25 Protein, His Tag
362.89€ - 2491.89€
Especificaciones
Número de acceso | NP_000408 |
---|---|
Peso molecular | 23.6 kDa |
Fuente | Human IL-2 R alpha, His Tag (ILA-H52H9) is expressed from human 293 cells (HEK293). It contains AA Glu 22 - Cys 213 (Accession # NP_000408). |
Inmunógeno | Glu 22 - Cys 213 |
Requisitos de almacenamiento | -20°C |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15897077
|
ACROBiosystems
ILA-H52H9-100UG |
100 μg |
362.89€
100 microgramos |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
15807087
|
ACROBiosystems
ILA-H52H9-1MG |
1 mg |
2491.89€
1 miligramo |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Description
Interleukin-2 receptor subunit alpha (IL2RA) is also known as IL-2R subunit alpha, IL-2-RA, IL2-RA, TAC antigen, p55, CD antigen CD25, is a type I transmembrane glycoprotein. IL2RA is expressed on activated T cells and regulatory T cells, and is capable of binding IL2 with low affinity by itself. However, a ligand-induced high affinity heterotrimeric receptor complex is produced when IL2RA is associated non-covelently with the IL2 receptor beta and gamma chain, and subsequently initiates the intacellular signal pathways such as MAPK or JAK/STAT. On dendritic cells (DC), CD25 has been previously regarded as an activation marker, while both murine and human DC can express CD25, they do not express the beta-chain of the IL-2 receptor, which is indispensable for the execution of IL-2 signaling.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 23.6 kDa. The protein migrates as 34-46 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
NP_000408 | |
Human IL-2 R alpha, His Tag (ILA-H52H9) is expressed from human 293 cells (HEK293). It contains AA Glu 22 - Cys 213 (Accession # NP_000408). | |
-20°C | |
IL-2 R alpha | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |
23.6 kDa | |
Glu 22 - Cys 213 | |
IL2RA,CD25,p55,IL2-RA,IL-2-RA | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |