Learn More
Invitrogen™ Human IL-1 alpha Recombinant Protein
Descripción
Mature human IL-1a was expressed in Pichia pastoris, KM71H. GenBank Accession # P01583, amino acids 113 - 271. For protein verification, the expressed protein was N-terminal sequenced and begins with the following 10 amino acids: SAPFSFLSNV. Purity is >95% by SDS-PAGE analysis. Activity: The ED50 typically falls into the range of 7.9 - 13.5 pg/mL in proliferation assays of mouse D10S cells. Use: For in vitro applications. A dose-response assay should be performed to determine the optimal concentration for specific experiments. Reconstitution and Dilution: Store lyophilized material at 4°C. Reconstitute in 0.5 mL of distilled or deionized water. Dilute in a buffer-containing carrier protein, aliquot, and store at or below -70°C, avoiding multiple freeze-thaw cycles. PBS with 1% BSA or a neutral media containing <10% FCS are suitable buffers.
Especificaciones
Especificaciones
| Número de acceso | P01583 |
| Para utilizar con (aplicación) | Control, ELISA, Immunoprecipitation, Western Blot |
| Formulación | PBS with no preservative |
| ID de gen (Entrez) | 3552 |
| Peso molecular | 18 kDa |
| Nombre | Human IL-1 alpha |
| Cantidad | 20 μg |
| Fuente | Yeast |
| Estado normativo | RUO |
| Alias de gen | CTLA8; hematopoietin-1; IL 1 A; IL 1 A; IL1; IL-1 alpha; IL17; IL1A; Il-1 A; IL-1 alpha; IL1-ALPHA; IL1F1; IL1a; ILN; Interleukin; interleukin 1 alpha; interleukin 1, alpha; interleukin 17; interleukin 1 A; interleukin 1-alpha; interleukin 1-alpha (AA 1 - 152); Interleukin1 alpha; interleukin-1 alpha; interleukin-1 alpha precursor; Interleukin-17 A; preinterleukin 1 alpha; pre-interleukin-1 alpha; pro-interleukin-1-alpha; RP23-160G19.8 |
| Mostrar más |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.