missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IRGC Synthetic Peptide
Synthetic Peptide
Marca: Invitrogen™ PEP0903
Detalles adicionales : Peso : 0.01000kg
Descripción
IRGC Synthetic Peptide, PEP-0903, from Invitrogen.
This peptide corresponds to 13 amino acids near the amino terminus of human IRGC.PEP-0903 can be used as a blocking peptide with polyclonal antibody PA5-20789.Especificaciones
56269 | |
purified | |
Human IRGC | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
Q6NXR0 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
IRGC | |
0.02% sodium azide | |
50 μg | |
PBS |