missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human IRX2 Full-length ORF (NP_150366.1, 1 a.a. - 471 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00153572-P01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16168763

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 04-09-2024
para ver el stock

Descripción

Descripción

IRX2 is a member of the Iroquois homeobox gene family. Members of this family appear to play multiple roles during pattern formation of vertebrate embryos.[supplied by OMIM]

Sequence: MSYPQGYLYQAPGSLALYSCPAYGASALAAPRSEELARSASGSAFSPYPGSAAFTAQAATGFGSPLQYSADAAAAAAGFPSYMGAPYDAHTTGMTGAISYHPYGSAAYPYQLNDPAYRKNATRDATATLKAWLNEHRKNPYPTKGEKIMLAIITKMTLTQVSTWFANARRRLKKENKMTWAPRNKSEDEDEDEGDATRSKDESPDKAQEGTETSAEDEGISLHVDSLTDHSCSAESDGEKLPCRAGDPLCESGSECKDKYDDLEDDEDDDEEGERGLAPPKPVTSSPLTGLEAPLLSPPPEAAPRGGRKTPQGSRTSPGAPPPASKPKLWSLAEIATSDLKQPSLGPGCGPPGLPAAAAPASTGAPPGGSPYPASPLLGRPLYYTSPFYGNYTNYGNLNAALQGQGLLRYNSAAAAPGEALHTAPKAASDAGKAGAHPLESHYRSPGGGYEPKKDASEGCTVVGGGVQPYL
Especificaciones

Especificaciones

NP_150366.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
75.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
IRXA2
IRX2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
153572
IRX2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSYPQGYLYQAPGSLALYSCPAYGASALAAPRSEELARSASGSAFSPYPGSAAFTAQAATGFGSPLQYSADAAAAAAGFPSYMGAPYDAHTTGMTGAISYHPYGSAAYPYQLNDPAYRKNATRDATATLKAWLNEHRKNPYPTKGEKIMLAIITKMTLTQVSTWFANARRRLKKENKMTWAPRNKSEDEDEDEGDATRSKDESPDKAQEGTETSAEDEGISLHVDSLTDHSCSAESDGEKLPCRAGDPLCESGSECKDKYDDLEDDEDDDEEGERGLAPPKPVTSSPLTGLEAPLLSPPPEAAPRGGRKTPQGSRTSPGAPPPASKPKLWSLAEIATSDLKQPSLGPGCGPPGLPAAAAPASTGAPPGGSPYPASPLLGRPLYYTSPFYGNYTNYGNLNAALQGQGLLRYNSAAAAPGEALHTAPKAASDAGKAGAHPLESHYRSPGGGYEPKKDASEGCTVVGGGVQPYL
RUO
IRX2
Wheat Germ (in vitro)
GST
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales