missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human KIAA2026 (aa 1844-1965) Control Fragment Recombinant Protein
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Descripción
Highest antigen sequence indentity to the following orthologs: Mouse (75%), Rat (75%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-51684 (PA5-51684. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Specifications
Specifications
| Número de acceso | Q5HYC2 |
| Concentración | ≥5.0 mg/mL |
| Para utilizar con (aplicación) | Blocking Assay, Control |
| Formulación | 1 M urea, PBS with no preservative; pH 7.4 |
| ID de gen (Entrez) | 158358 |
| Nombre | Human KIAA2026 (aa 1844-1965) Control Fragment |
| Cantidad | 100 μL |
| Estado normativo | RUO |
| Alias de gen | KIAA2026; uncharacterized protein KIAA2026 |
| Nombre común | KIAA2026 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction