Learn More
ACROBiosystems Human LAIR1 / CD305 Protein, Fc Tag
423.38€ - 2951.55€
Especificaciones
Número de acceso | AAH27899.1 |
---|---|
Peso molecular | 42.1 kDa |
Fuente | Human LAIR-1, Fc Tag (LA1-H5252) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - His 163 (Accession # Q6GT x 8-1). |
Inmunógeno | Gln 22 - His 163 |
Requisitos de almacenamiento | -20°C |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15897197
|
ACROBiosystems
LA1-H5252-1MG |
1 mg |
2951.55€
1 miligramo |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
15887197
|
ACROBiosystems
LA1-H5252-100UG |
100 μg |
423.38€
100 microgramos |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
Leukocyte-associated immunoglobulin-like receptor-1 (LAIR-1) is constitutively expressed on the majority of human peripheral blood mononuclear leukocytes. LAIR-1 or CD305 is a transmembrane glycoprotein with a single immunoglobulin-like domain and a cytoplasmic tail containing two immune receptor tyrosine-based inhibitory motifs. LAIR-1 recruits SHP-1 and SHP-2 phosphatases upon activation, and cross-linking of the LAIR-1 antigen on natural killer (NK) cells results in strong inhibition of NK cell–mediated cytotoxicity. Functions as an inhibitory receptor that plays a constitutive negative regulatory role on cytolytic function of natural killer (NK) cells, B-cells and T-cells. Activation by Tyr phosphorylation results in recruitment and activation of the phosphatases PTPN6 and PTPN11. It also reduces the increase of intracellular calcium evoked by B-cell receptor ligation.Diseases associated with LAIR1 include Chronic Active Epstein-Barr Virus Infection and Palindromic Rheumatism.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 42.1 kDa. The protein migrates as 55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
AAH27899.1 | |
Human LAIR-1, Fc Tag (LA1-H5252) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - His 163 (Accession # Q6GT x 8-1). | |
-20°C | |
LAIR-1 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Immune Checkpoint Proteins |
42.1 kDa | |
Gln 22 - His 163 | |
LAIR1,CD305 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |