missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human LSECtin / CLEC4G Protein, Fc Tag
350.80€ - 2419.31€
Especificaciones
Número de acceso | NP_940894.1 |
---|---|
Peso molecular | 52.1 kDa |
Fuente | Human LSECtin, Fc Tag (CLG-H5250) is expressed from human 293 cells (HEK293). It contains AA Lys 55 - Cys 293 (Accession # Q6UXB4-1). |
Inmunógeno | Lys 55 - Cys 293 |
Requisitos de almacenamiento | -20°C |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15886447
|
ACROBiosystems
CLG-H5250-1MG |
1 mg |
2419.31€
1 miligramo |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
15896447
|
ACROBiosystems
CLG-H5250-50UG |
50 μg |
350.80€
50 microgramos |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
C-type lectin superfamily 4, member G (CLEC4G) is also known as liver and lymph node sinusoidal endothelial cell C-type lectin (LSECtin), which is a member of subgroup II of the C-type (Ca2+-dependent) lectin superfamily. CLEC4G / LSECtin binds to mannose, GlcNAc, and fucose in a Ca2+-dependent manner. In addition, CLEC4G / LSECtin has the ability to bind to surface glycoproteins of enveloped viruses. In particular, interaction of LSECtin with the surface glycoproteins of severe acute respiratory syndrome (SARS) coronavirus and Ebola virus has been described, and LSECtin-mediated infection of cells by Ebola virus has been demonstrated.
This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 52.1 kDa. The protein migrates as 58-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
NP_940894.1 | |
Human LSECtin, Fc Tag (CLG-H5250) is expressed from human 293 cells (HEK293). It contains AA Lys 55 - Cys 293 (Accession # Q6UXB4-1). | |
-20°C | |
LSECtin | |
Human | |
50 mM Tris, 100 mM Glycine, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |
52.1 kDa | |
Lys 55 - Cys 293 | |
CLEC4G,LSECtin | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>85% as determined by SDS-PAGE |