Learn More
ACROBiosystems Human LYPD3 / C4.4A Protein, His Tag
Marca: ACROBiosystems LY3-H52H5-100UG
Detalles adicionales : Peso : 0.00001kg
Descripción
Ly6/PLAUR domain-containing protein 3 (LYPD3) is also known as MIG-C4 ,C4.4A, GPI-anchored metastasis-associated protein C4.4A homolog and Matrigel-induced gene C4 protein,which is a cell membrane protein containing two UPAR/Ly6 domains. LYPD3 is expressed in placenta, skin and urothelium. LYPD3 is found in the majority of primary and metastatic transitional cell carcinomas (TCCs) and as well in breast cancer tissues, but not in adjacent normal tissues. LYPD3 play a role in supporting cell migration and it is not only probably involved in urothelial cell-matrix interactions, but also in tumor progression.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28.7 kDa. The protein migrates as 45-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
AAH39167 | |
100 μg | |
Leu 31 - His 286 | |
LYPD3,MIG-C4,C4.4 A | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
28.7 kDa | |
Human LYPD3, His Tag (LY3-H52H5) is expressed from human 293 cells (HEK293). It contains AA Leu 31 - His 286 (Accession # AAH39167). | |
-20°C | |
LYPD3 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |