Learn More
ACROBiosystems Human MMP-9 Protein, His Tag
423.38€ - 2903.17€
Especificaciones
Número de acceso | AAH06093 |
---|---|
Peso molecular | 50.8 kDa |
Fuente | Human MMP-9, His Tag (MM9-H5221) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Pro 469 (Accession # AAH06093). |
Inmunógeno | Ala 20 - Pro 469 |
Requisitos de almacenamiento | -20°C |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15897307
|
ACROBiosystems
MM9-H5221-50UG |
50 μg |
423.38€
50 microgramos |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
15887307
|
ACROBiosystems
MM9-H5221-1MG |
1 mg |
2903.17€
1 miligramo |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
Matrix metallopeptidase 9 (MMP-9) is also known as 92 kDa type IV collagenase, 92 kDa gelatinase or gelatinase B (GELB), CLG4B, is secreted from neutrophils, macrophages, and a number of transformed cells, and is the most complex family member in terms of domain structure and regulation of its activity. . Structurally, MMP9 maybe be divided into five distinct domains: a prodomain which is cleaved upon activation, a gelatinbinding domain consisting of three contiguous fibronectin type II units, a catalytic domain containing the zinc binding site, a prolinerich linker region, and a carboxyl terminal hemopexinlike domain. This enzyme degrades various substrates including gelatin, collagen types IV and V, and elastin. MMP9 is involved in a variety of autoimmune diseases such as systemic lupus erythematosus, rheumatoid arthritis, and multiple sclerosis, and be regarded as a potential therapeutic target.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 50.8 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
AAH06093 | |
Human MMP-9, His Tag (MM9-H5221) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Pro 469 (Accession # AAH06093). | |
-20°C | |
MMP-9 | |
Human | |
50 mM Tris, 150 mM NaCl, pH7.5 | |
HEK293 | |
Enzymes |
50.8 kDa | |
Ala 20 - Pro 469 | |
MMP9,CLG4B,GELB,MANDP2,Gelatinase B | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |