missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Gelsolin/GSN Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 1 publication
Marca: R&D Systems MAB8170
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Gelsolin/GSN Monoclonal specifically detects Gelsolin/GSN in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.Especificaciones
Gelsolin/GSN | |
Monoclonal | |
LYOPH | |
Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Immunohistochemistry 8-25 ug/mL | |
Actin-depolymerizing factor, ADF, AGEL, Brevin, DKFZp313L0718, gelsolin, gelsolin (amyloidosis, Finnish type), GSN | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human Gelsolin/GSN in ELISAs. Detects human, mouse and rat Gelsolin/GSN in Western Blots. | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry | |
893205 | |
Unconjugated | |
P06396 | |
GSN | |
HEK293 human embryonic kidney cell line transfected with human Gelsolin/GSN Met1-Ala782 Accession # P06396 | |
100 μg | |
2934 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |