missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Serpin F1/PEDF Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 8 publications
Marca: R&D Systems AF1177
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Serpin F1/PEDF Polyclonal specifically detects Serpin F1/PEDF in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.Especificaciones
Serpin F1/PEDF | |
Polyclonal | |
Unconjugated | |
P36955 | |
SERPINF1 | |
E. coli-derived recombinant human Serpin F1/PEDF Asp44-Pro418 Accession # P36955 | |
100 μg | |
Primary | |
Detects human Serpin F1/PEDF in direct ELISAs. Detects human and mouse Serpin F1/PEDF in Western blots. | |
Human, Mouse | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.2-2 ug/mL, Simple Western 2 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Cell proliferation-inducing gene 35 protein, EPC-1, EPC-1PIG35, PEDF, PEDFpigment epithelium-derived factor, pigment epithelium derived factor), member 1, proliferation-inducing protein 35, serine (or cysteine) proteinase inhibitor, clade F (alpha-2 antiplasmin, Serpin F1, serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epitheliumderived factor), member 1 | |
Goat | |
Affinity Purified | |
RUO | |
5176 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido