Learn More
ACROBiosystems Human NTB-A / SLAMF6 Protein, His Tag
Marca: ACROBiosystems NTA-H52HA-100UG
Detalles adicionales : Peso : 0.00001kg
Descripción
The SLAM family of receptors is composed of six members named SLAM (CD150); 2B4 (CD244); Ly-9 (CD229); CD84; natural killer, T and B cell antigen (NTB-A; Ly108 in the mouse. Most, if not all,immune cell types express more than one SLAM family receptor. All six SLAM-related genes are located within a 400–500 kilobase (kb) genomic segment on chromosome 1 in humans and mice. This feature implies that the genes that encode the SLAM family were created by serial gene duplication.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.0 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
NP_001171643.1 | |
100 μg | |
Gln 22 - Met 226 | |
NTB-A,SLAMF6,Ly108,NK-T-B-antigen,CD352,KALI | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
25.0 kDa | |
Human NTB-A, His Tag (NTA-H52Ha) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Met 226 (Accession # Q96DU3-1). | |
-20°C | |
NTB-A | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |