missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human OPRK1 Partial ORF (NP_000903, 1 a.a. - 58 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00004986-Q01.25ug
Additional Details : Peso : 0.00010kg
Description
Sequence: MDSPIQIFRGEPGPTCAPSACLPPNSSAWFPGWAEPDSNGSAGSEDAQLEPAHISPAISpecifications
NP_000903 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.12kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MDSPIQIFRGEPGPTCAPSACLPPNSSAWFPGWAEPDSNGSAGSEDAQLEPAHISPAI | |
RUO | |
OPRK1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
4986 | |
OPRK1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KOR/OPRK | |
OPRK1 | |
Recombinant | |
wheat germ expression system |