Learn More
ACROBiosystems Human PD-L1 / B7-H1 Protein, Fc Tag
MALS verified
Marca:  ACROBiosystems PD1-H5258-100UG
Detalles adicionales : Peso : 0.00001kg
 Ver más versiones de este producto
Código de producto. 15897457
  • 387.09€ / 100 microgramos
Fecha estimada de envío: 07-08-2024
para ver el stock
Descripción
Descripción
Programmed cell death 1 ligand 1 (PD-L1) is also known as cluster of differentiation (CD274) or B7 homolog 1 (B7-H1), is a member of the growing B7 family of immune molecules and is involved in the regulation of cellular and humoral immune responses. B7-H1 is a cell surface immunoglobulin superfamily with two Ig-like domains within the extracellular region and a short cytoplasmic domain. PD-L1 is highly expressed in the heart, skeletal muscle, placenta and lung and weakly expressed in the thymus, spleen, kidney and liver. PD-L1 is expressed on activated T-cells, B-cells, dendritic cells, keratinocytes and monocytes. PD-L1 is up-regulated on T- and B-cells, dendritic cells, keratinocytes and monocytes after LPS and IFNG activation and up-regulated in B-cells activated by surface Ig cross-linking. PD-L1 involve in the costimulatory signal, essential for T-cell proliferation and production of IL10 and IFNG, in an IL2-dependent and a PDCD1-independent manner.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 51.3 kDa. The protein migrates as 60-75 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Especificaciones
Especificaciones
NP_054862.1
100 μg
Phe 19 - Arg 238
PD-L1,CD274,B7-H1,PDCD1L1,PDCD1LG1
Unconjugated
Fc Tag
Less than 0.1 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
51.3 kDa
Human PD-L1, Fc Tag (PD1-H5258) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Arg 238 (Accession # NP_054862.1).
-20°C
PD-L1
Human
PBS, pH7.4
HEK293
Immune Checkpoint Proteins