Learn More
Human Protocadherin alpha 1 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Marca:  R&D Systems AF5064
Código de producto. 18052982
  • 464.80€ / 100 microgramos
Explore más ofertas especiales
Descripción
Descripción
Protocadherin alpha 1 Polyclonal specifically detects Protocadherin alpha 1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Especificaciones
Especificaciones
Protocadherin alpha 1
Polyclonal
Unconjugated
Q9Y5I3
PCDHA1
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Protocadherin alpha 1 Gln30-Pro689 Accession # Q9Y5I3
100 μg
Primary
Detects human Protocadherin alpha 1 (PCDH alpha 1) in direct ELISAs and Western blots.
Human
IgG
Western Blot, Immunohistochemistry
LYOPH
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL
KIAA0345-like 13, PCDHA1, PCDH-alpha-1, PCDH-ALPHA1, protocadherin alpha 1, protocadherin alpha-1
Sheep
Affinity Purified
RUO
56147
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.