Learn More
ACROBiosystems Human PTPRD Protein, His Tag
508.05€ - 3362.84€
Especificaciones
Número de acceso | NP_002830.1 |
---|---|
Peso molecular | 140.0 kDa |
Fuente | Human PTPRD, His Tag (PTD-H52H9) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Glu 1265 (Accession # P23468-1). |
Inmunógeno | Glu 21 - Glu 1265 |
Requisitos de almacenamiento | -20°C |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15897557
|
ACROBiosystems
PTD-H52H9-100UG |
100 μg |
508.05€
100 microgramos |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
15807567
|
ACROBiosystems
PTD-H52H9-1MG |
1 mg |
3362.84€
1 miligramo |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
Receptor-type tyrosine-protein phosphatase delta is an enzyme that is encoded by the PTPRD gene. The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. PTPRD has been shown to interact with PTPRS and liprin-alpha-1. Studies of the similar genes in chick and fly suggest the role of this PTP is in promoting neurite growth, and regulating neurons axon guidance. Multiple tissue specific alternatively spliced transcript variants of this gene have been reported. Mutations in the PTPRD gene are associated with autism, obsessive–compulsive disorder, and breast cancer.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 140.0 kDa. The protein migrates as 19 kDa and 120-130 kDa under reducing (R) condition (SDS-PAGE).Especificaciones
NP_002830.1 | |
Human PTPRD, His Tag (PTD-H52H9) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Glu 1265 (Accession # P23468-1). | |
-20°C | |
PTPRD | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Enzymes |
140.0 kDa | |
Glu 21 - Glu 1265 | |
PTPRD,R-PTP-delta | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |