Learn More
STEMCELL Technologies Human Recombinant IL-33
Código de producto. p-7217545
Encompass_Preferred
Click to view available options
Cantidad:
100 μg
10 μg
Tamaño de la unidad:
10 microgramos
100 microgramos
Código de producto. 17189421
Marca: STEMCELL Technologies 78043
Interleukin 33
  • Interleukin 33 (IL-33) is a pro-inflammatory cytokine from the IL-1 family
  • It binds to ST2 receptor and activates NF-κB and MAPK pathways
  • IL-33 is expressed by epithelial cells, smooth muscle cells, and fibroblasts in various tissues and organs, as well as resting basophils, mast cells, eosinophils, natural helper cells, group 2 innate lymphoid cells, dendritic cells, and activated macrophages (Schmitz et al.; Yasuda et al.) It contributes to allergic inflammation by stimulating production of the cytokines IL-4, IL-5, and IL-13, and stimulates host defense against microbial and viral infections (Liew; Yasuda et al.) In the central nervous system, IL-33 is produced by endothelial cells and astrocytes
  • It induces proliferation of microglia and mediates production of pro-inflammatory cytokines (Yasuoka et al.)
Especificaciones
Nombre IL-1F11, Interleukin-33, NP-HEV
Cantidad 100 μg
Tipo de célula Monocytes; T Cells, Other Subsets
Grado de pureza o calidad ≥95%