Learn More
STEMCELL Technologies Human Recombinant TNF-beta
Código de producto. 17159621
Encompass_Preferred
Click to view available options
Cantidad:
10 μg
50 μg
Tamaño de la unidad:
10 microgramos
50 microgramos
Código de producto. 17159621
Marca: STEMCELL Technologies 78125.1
Tumor necrosis factor beta
  • Tumor necrosis factor-beta (TNF-β), also known as lymphotoxin-alpha, is a member of the TNF family
  • It is a pro-inflammatory cytokine that activates NF-kB, MAPK, and PI3K/AKT pathways upon binding to TNF receptors 1 and 2
  • It is produced mainly by T cells, though other cells can express TNF-β at lower levels (Chu)
  • TNF-β is involved in autoimmune disorders, lymph node development, and mediating the inflammatory demyelination process (Seleznik et al.; McCarthy et al.)
Especificaciones
Nombre LT-α, Lymphotoxin-alpha, TNF-beta, TNFSF1, Tumor necrosis factor-β
Cantidad 50 μg
Tipo de célula T Cells
Grado de pureza o calidad ≥95%