Learn More
ACROBiosystems Human ROR1 (308-395, Kringle domain) Protein, His Tag
495.95€ - 3762.02€
Especificaciones
Número de acceso | Q01973 |
---|---|
Peso molecular | 11.2 kDa |
Fuente | Human ROR1 (308-395, Kringle domain), His Tag (RO1-H5223) is expressed from human 293 cells (HEK293). It contains AA Asn 308 - Asp 395 (Accession # Q01973-1). |
Inmunógeno | Asn 308 - Asp 395 |
Requisitos de almacenamiento | -20°C |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15887587
|
ACROBiosystems
RO1-H5223-50UG |
50 μg |
495.95€
50 microgramos |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
15877587
|
ACROBiosystems
RO1-H5223-1MG |
1 mg |
3762.02€
1 miligramo |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
Tyrosine-protein kinase transmembrane receptor ROR1 is also known as Neurotrophic tyrosine kinase, receptor-related 1 (NTRKR1), which belongs to the protein kinase superfamily or tyr protein kinase family or ROR subfamily. ROR1 contains 1 FZ (frizzled) domain, 1 Ig-like C2-type (immunoglobulin-like) domain, 1 kringle domain, 1 protein kinase domain. ROR1 is expressed at high levels during early embryonic development. The expression levels drop strongly around day 16 and there are only very low levels in adult tissues. Isoform Short is strongly expressed in fetal and adult CNS and in a variety of human cancers, including those originating from CNS or PNS neuroectoderm. ROR1 could interact with casein kinase 1 epsilon (CK1ε) to activate phosphoinositide 3-kinase-mediated AKT phosphorylation and cAMP-response-element-binding protein (CREB), which was associated with enhanced tumor-cell growth.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 11.2 kDa. The protein migrates as 16-18 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
Q01973 | |
Human ROR1 (308-395, Kringle domain), His Tag (RO1-H5223) is expressed from human 293 cells (HEK293). It contains AA Asn 308 - Asp 395 (Accession # Q01973-1). | |
-20°C | |
ROR1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |
11.2 kDa | |
Asn 308 - Asp 395 | |
ROR1,NTRKR1 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |